Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289010.1 | NP_001275939.1 | 1 - 2 | 872 | 95 |
NM_020379.3 | NP_065112.1 | 1 - 2 | 872 | 95 | |
XM_017001860.1 | XP_016857349.1 | 1 - 2 | 91 | 95 | |
XM_017001861.1 | XP_016857350.1 | 1 - 2 | 1520 | 95 | |
GenBank mRNA | AB209275.1 | - | 1 - 2 | 220 | 95 |
AF261655.1 | - | 1 - 2 | 872 | 95 | |
BC137017.1 | - | 1 - 2 | 678 | 95 |