Assay Design



Gene Transcripts

Gene Symbol
KIFC2
Entrez Gene ID
90990
Gene Name
kinesin family member C2
Gene Aliases
-
Chromosome Location
Chr.8: 144466246 - 144474116 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005272357.3 XP_005272414.1 17 - 18 2292 73
GenBank mRNA AK055045.1 - 12 - 13 1375 73

View Full Product Details