Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040118.2 | NP_001035207.1 | 16 - 17 | 2491 | 73 |
NM_001135190.1 | NP_001128662.1 | 13 - 14 | 2119 | 73 | |
NM_015242.4 | NP_056057.2 | 14 - 15 | 2302 | 73 | |
GenBank mRNA | AB018325.2 | - | 14 - 15 | 2279 | 73 |
AF411983.1 | - | 10 - 11 | 1360 | 73 | |
AJ621557.1 | - | 14 - 15 | 2302 | 73 | |
AK160383.1 | - | 13 - 14 | 2640 | 73 | |
AY049732.1 | - | 11 - 12 | 2139 | 73 | |
AY553630.1 | - | 16 - 17 | 2506 | 73 | |
BC140792.1 | - | 13 - 14 | 1789 | 73 | |
BC144562.1 | - | 14 - 15 | 1972 | 73 |