Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040118.2 | NP_001035207.1 | 25 - 26 | 3700 | 91 |
NM_001135190.1 | NP_001128662.1 | 22 - 23 | 3328 | 91 | |
NM_015242.4 | NP_056057.2 | 23 - 24 | 3511 | 91 | |
GenBank mRNA | AB018325.2 | - | 23 - 24 | 3488 | 91 |
AF411983.1 | - | 19 - 20 | 2569 | 91 | |
AJ621557.1 | - | 23 - 24 | 3511 | 91 | |
AK160383.1 | - | 21 - 22 | 4178 | 91 | |
AY049732.1 | - | 20 - 21 | 3348 | 91 | |
AY553630.1 | - | 25 - 26 | 3715 | 91 | |
BC008315.1 | - | 7 - 8 | 981 | 91 | |
BC056401.1 | - | 11 - 12 | 1354 | 91 | |
BC140792.1 | - | 22 - 23 | 2998 | 91 | |
BC144562.1 | - | 23 - 24 | 3181 | 91 |