Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005819.5 | NP_005810.1 | 4 - 5 | 816 | 125 |
XM_011509058.1 | XP_011507360.1 | 4 - 5 | 816 | 125 | |
GenBank mRNA | AJ002078.1 | - | 4 - 5 | 368 | 125 |
AK312440.1 | - | 4 - 5 | 603 | 125 | |
BC009944.2 | - | 4 - 5 | 539 | 125 | |
BU902578.1 | - | 4 - 5 | 606 | 125 | |
CR541856.1 | - | 4 - 5 | 368 | 125 | |
CR541873.1 | - | 4 - 5 | 368 | 125 |