Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012387.2 | NP_036519.2 | 10 - 11 | 1186 | 96 |
XM_011541150.1 | XP_011539452.1 | 8 - 9 | 998 | 96 | |
XM_011541152.1 | XP_011539454.1 | 7 - 8 | 718 | 96 | |
XM_011541157.1 | XP_011539459.1 | 7 - 8 | 665 | 96 | |
GenBank mRNA | AB017919.1 | - | 10 - 11 | 96 | |
AK290507.1 | - | 10 - 11 | 1184 | 96 | |
AK314839.1 | - | 10 - 11 | 1186 | 96 | |
BC025718.1 | - | 10 - 11 | 96 |