Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321120.1 | NP_001308049.1 | 7 - 8 | 957 | 127 |
NM_018488.3 | NP_060958.2 | 6 - 7 | 797 | 127 | |
XM_011525490.2 | XP_011523792.1 | 7 - 8 | 995 | 127 | |
XM_011525491.2 | XP_011523793.1 | 7 - 8 | 995 | 127 | |
XM_011525495.2 | XP_011523797.1 | 7 - 8 | 998 | 127 | |
GenBank mRNA | AF188703.1 | - | 6 - 7 | 839 | 127 |
AK310312.1 | - | 3 - 4 | 324 | 127 | |
BC136403.1 | - | 7 - 8 | 821 | 127 | |
BC142620.1 | - | 7 - 8 | 957 | 127 | |
BC144062.1 | - | 7 - 8 | 821 | 127 |