Assay Design



Gene Transcripts

Gene Symbol
KRT12
Entrez Gene ID
3859
Gene Name
keratin 12
Gene Aliases
K12
Chromosome Location
Chr.17: 40861178 - 40867210 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000223.3 NP_000214.1 4 - 5 994 64
GenBank mRNA AK313747.1 - 4 - 5 1008 64
D78367.1 - 4 - 5 994 64

View Full Product Details