Assay Design



Gene Transcripts

Gene Symbol
AMTN
Entrez Gene ID
401138
Gene Name
amelotin
Gene Aliases
UNQ689
Chromosome Location
Chr.4: 70518572 - 70532743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286731.1 NP_001273660.1 6 - 7 430 80
NM_212557.3 NP_997722.1 6 - 7 433 80
GenBank mRNA AB593161.1 - 6 - 7 432 80
AY358528.1 - 6 - 7 424 80
BC121812.1 - 6 - 7 401 80
BC121817.1 - 6 - 7 404 80

View Full Product Details