Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000833.4 | NP_000824.1 | 6 - 7 | 1637 | 62 |
NM_001134407.2 | NP_001127879.1 | 5 - 6 | 1875 | 62 | |
NM_001134408.2 | NP_001127880.1 | 5 - 6 | 1536 | 62 | |
XM_011522458.2 | XP_011520760.1 | 4 - 5 | 1123 | 62 | |
XM_011522461.2 | XP_011520763.1 | 6 - 7 | 2247 | 62 | |
XM_017023172.1 | XP_016878661.1 | 4 - 5 | 1748 | 62 | |
XM_017023173.1 | XP_016878662.1 | 4 - 5 | 1748 | 62 | |
GenBank mRNA | AB209695.1 | - | 5 - 6 | 1875 | 62 |
AK295620.1 | - | 4 - 5 | 968 | 62 | |
BC117131.1 | - | 4 - 5 | 1356 | 62 | |
BC143273.1 | - | 4 - 5 | 1356 | 62 | |
U09002.1 | - | 5 - 6 | 1482 | 62 | |
U90277.1 | - | 6 - 7 | 1637 | 62 |