Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202523.1 | NP_001189452.1 | 12 - 13 | 2053 | 74 |
NM_001297652.1 | NP_001284581.1 | 12 - 13 | 2172 | 74 | |
NM_001297653.1 | NP_001284582.1 | 12 - 13 | 1974 | 74 | |
NM_001297654.1 | NP_001284583.1 | 13 - 14 | 2085 | 74 | |
NM_001954.4 | NP_001945.3 | 14 - 15 | 2092 | 74 | |
NM_013993.2 | NP_054699.2 | 14 - 15 | 2129 | 74 | |
NM_013994.2 | NP_054700.2 | 12 - 13 | 1912 | 74 | |
XM_006715185.2 | XP_006715248.1 | 13 - 14 | 2076 | 74 | |
XM_011514882.2 | XP_011513184.1 | 13 - 14 | 2076 | 74 | |
XM_011514883.1 | XP_011513185.1 | 15 - 16 | 2191 | 74 | |
XM_011514884.1 | XP_011513186.1 | 15 - 16 | 2267 | 74 | |
XM_011514885.1 | XP_011513187.1 | 13 - 14 | 2114 | 74 | |
XM_011514886.1 | XP_011513188.1 | 13 - 14 | 1979 | 74 | |
XM_011514887.1 | XP_011513189.1 | 13 - 14 | 2129 | 74 | |
XM_011514888.2 | XP_011513190.1 | 12 - 13 | 1965 | 74 | |
XM_017011268.1 | XP_016866757.1 | 13 - 14 | 1946 | 74 | |
XM_017011269.1 | XP_016866758.1 | 12 - 13 | 2018 | 74 | |
GenBank mRNA | AB210021.1 | - | 13 - 14 | 1913 | 74 |
AK130776.1 | - | 8 - 9 | 1066 | 74 | |
AK291621.1 | - | 12 - 13 | 1867 | 74 | |
AK294793.1 | - | 12 - 13 | 2039 | 74 | |
BC008716.2 | - | 14 - 15 | 2092 | 74 | |
BC013400.2 | - | 14 - 15 | 2092 | 74 | |
BC070070.1 | - | 12 - 13 | 1863 | 74 | |
L11315.1 | - | 14 - 15 | 2011 | 74 | |
L20817.1 | - | 13 - 14 | 2062 | 74 | |
L57508.1 | - | 12 - 13 | 1951 | 74 | |
X74979.1 | - | 11 - 12 | 1804 | 74 | |
Z29093.1 | - | 11 - 12 | 2095 | 74 |