Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202521.1 | NP_001189450.1 | 12 - 13 | 1899 | 102 |
NM_001202522.1 | NP_001189451.1 | 12 - 13 | 1817 | 102 | |
NM_001202523.1 | NP_001189452.1 | 14 - 15 | 2396 | 102 | |
NM_001297652.1 | NP_001284581.1 | 14 - 15 | 2515 | 102 | |
NM_001297653.1 | NP_001284582.1 | 14 - 15 | 2317 | 102 | |
NM_001297654.1 | NP_001284583.1 | 15 - 16 | 2428 | 102 | |
NM_001954.4 | NP_001945.3 | 16 - 17 | 2435 | 102 | |
NM_013993.2 | NP_054699.2 | 16 - 17 | 2472 | 102 | |
NM_013994.2 | NP_054700.2 | 14 - 15 | 2273 | 102 | |
XM_006715185.2 | XP_006715248.1 | 15 - 16 | 2419 | 102 | |
XM_011514882.2 | XP_011513184.1 | 15 - 16 | 2437 | 102 | |
XM_011514883.1 | XP_011513185.1 | 17 - 18 | 2552 | 102 | |
XM_011514884.1 | XP_011513186.1 | 17 - 18 | 2628 | 102 | |
XM_011514885.1 | XP_011513187.1 | 15 - 16 | 2475 | 102 | |
XM_011514886.1 | XP_011513188.1 | 15 - 16 | 2340 | 102 | |
XM_011514887.1 | XP_011513189.1 | 15 - 16 | 2490 | 102 | |
XM_011514888.2 | XP_011513190.1 | 14 - 15 | 2326 | 102 | |
XM_017011268.1 | XP_016866757.1 | 15 - 16 | 2307 | 102 | |
XM_017011269.1 | XP_016866758.1 | 14 - 15 | 2379 | 102 | |
GenBank mRNA | AB210021.1 | - | 15 - 16 | 2256 | 102 |
AK130776.1 | - | 10 - 11 | 1409 | 102 | |
AK291621.1 | - | 14 - 15 | 2210 | 102 | |
AK294793.1 | - | 14 - 15 | 2382 | 102 | |
BC008716.2 | - | 16 - 17 | 2435 | 102 | |
BC013400.2 | - | 16 - 17 | 2435 | 102 | |
BI193181.1 | - | 2 - 3 | 316 | 102 | |
L11315.1 | - | 16 - 17 | 2354 | 102 | |
L20817.1 | - | 15 - 16 | 2405 | 102 | |
L57508.1 | - | 14 - 15 | 2294 | 102 | |
X74979.1 | - | 13 - 14 | 2147 | 102 | |
Z29093.1 | - | 13 - 14 | 2438 | 102 |