Assay Design
Gene Transcripts
- Gene Symbol
- DDR1
- Entrez Gene ID
-
780
- Gene Name
- discoidin domain receptor tyrosine kinase 1
- Gene Aliases
-
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
- Chromosome Location
-
Chr.6: 30880909 - 30900156 on Build GRCh38
View Full Product Details