Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003004.2 | NP_002995.1 | 2 - 3 | 490 | 83 |
XM_005256392.3 | XP_005256449.1 | 2 - 3 | 274 | 83 | |
XM_011523588.2 | XP_011521890.1 | 2 - 3 | 521 | 83 | |
GenBank mRNA | AK223068.1 | - | 2 - 3 | 183 | 83 |
AK223104.1 | - | 2 - 3 | 214 | 83 | |
AK290708.1 | - | 2 - 3 | 443 | 83 | |
AK312980.1 | - | 2 - 3 | 210 | 83 | |
BC017716.1 | - | 2 - 3 | 184 | 83 | |
U77643.1 | - | 2 - 3 | 210 | 83 |