Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 21 - 22 | 3507 | 69 |
XM_005251773.2 | XP_005251830.1 | 21 - 22 | 3416 | 69 | |
XM_005251776.2 | XP_005251833.1 | 20 - 21 | 3256 | 69 | |
XM_011518339.2 | XP_011516641.1 | 22 - 23 | 3491 | 69 | |
XM_011518340.2 | XP_011516642.1 | 23 - 24 | 3706 | 69 | |
XM_011518341.2 | XP_011516643.1 | 22 - 23 | 3491 | 69 | |
XM_011518342.2 | XP_011516644.1 | 19 - 20 | 3194 | 69 | |
XM_011518344.2 | XP_011516646.1 | 22 - 23 | 3491 | 69 | |
XM_017014378.1 | XP_016869867.1 | 22 - 23 | 3689 | 69 | |
XM_017014379.1 | XP_016869868.1 | 22 - 23 | 4097 | 69 | |
XM_017014380.1 | XP_016869869.1 | 22 - 23 | 3423 | 69 | |
XM_017014381.1 | XP_016869870.1 | 22 - 23 | 3489 | 69 | |
XM_017014382.1 | XP_016869871.1 | 20 - 21 | 3954 | 69 | |
GenBank mRNA | AB055982.1 | - | 20 - 21 | 3103 | 69 |
AB445477.1 | - | 20 - 21 | 3103 | 69 | |
AF165281.1 | - | 19 - 20 | 3043 | 69 | |
AF285167.1 | - | 21 - 22 | 3393 | 69 | |
AJ012376.1 | - | 19 - 20 | 3043 | 69 | |
BC146856.1 | - | 21 - 22 | 3317 | 69 |