Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 34 - 35 | 5102 | 103 |
XM_005251773.2 | XP_005251830.1 | 34 - 35 | 5017 | 103 | |
XM_005251776.2 | XP_005251833.1 | 33 - 34 | 4857 | 103 | |
XM_011518339.2 | XP_011516641.1 | 35 - 36 | 5092 | 103 | |
XM_011518340.2 | XP_011516642.1 | 36 - 37 | 5307 | 103 | |
XM_011518341.2 | XP_011516643.1 | 35 - 36 | 5086 | 103 | |
XM_011518342.2 | XP_011516644.1 | 32 - 33 | 4795 | 103 | |
XM_017014378.1 | XP_016869867.1 | 35 - 36 | 5290 | 103 | |
XM_017014379.1 | XP_016869868.1 | 35 - 36 | 5698 | 103 | |
XM_017014380.1 | XP_016869869.1 | 35 - 36 | 5024 | 103 | |
XM_017014381.1 | XP_016869870.1 | 35 - 36 | 5090 | 103 | |
XM_017014382.1 | XP_016869871.1 | 33 - 34 | 5555 | 103 | |
GenBank mRNA | AB055982.1 | - | 33 - 34 | 4698 | 103 |
AB445477.1 | - | 33 - 34 | 4698 | 103 | |
AF165281.1 | - | 32 - 33 | 4638 | 103 | |
AF285167.1 | - | 34 - 35 | 4988 | 103 | |
AJ012376.1 | - | 32 - 33 | 4638 | 103 | |
BC146856.1 | - | 34 - 35 | 4912 | 103 |