Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 41 - 42 | 6045 | 111 |
XM_005251773.2 | XP_005251830.1 | 41 - 42 | 5960 | 111 | |
XM_005251776.2 | XP_005251833.1 | 40 - 41 | 5800 | 111 | |
XM_011518339.2 | XP_011516641.1 | 42 - 43 | 6035 | 111 | |
XM_011518340.2 | XP_011516642.1 | 43 - 44 | 6250 | 111 | |
XM_011518341.2 | XP_011516643.1 | 42 - 43 | 6029 | 111 | |
XM_011518342.2 | XP_011516644.1 | 39 - 40 | 5738 | 111 | |
XM_017014378.1 | XP_016869867.1 | 42 - 43 | 6233 | 111 | |
XM_017014379.1 | XP_016869868.1 | 42 - 43 | 6641 | 111 | |
XM_017014380.1 | XP_016869869.1 | 42 - 43 | 5967 | 111 | |
XM_017014381.1 | XP_016869870.1 | 42 - 43 | 6033 | 111 | |
XM_017014382.1 | XP_016869871.1 | 40 - 41 | 6498 | 111 | |
GenBank mRNA | AB055982.1 | - | 40 - 41 | 5641 | 111 |
AB208839.1 | - | 5 - 6 | 597 | 111 | |
AB445477.1 | - | 40 - 41 | 5641 | 111 | |
AF165281.1 | - | 39 - 40 | 5581 | 111 | |
AF285167.1 | - | 41 - 42 | 5931 | 111 | |
AJ012376.1 | - | 39 - 40 | 5581 | 111 | |
AK027864.1 | - | 6 - 7 | 768 | 111 | |
BC146856.1 | - | 41 - 42 | 5855 | 111 |