Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 6 - 7 | 947 | 80 |
XM_005251773.2 | XP_005251830.1 | 6 - 7 | 856 | 80 | |
XM_005251776.2 | XP_005251833.1 | 5 - 6 | 696 | 80 | |
XM_011518339.2 | XP_011516641.1 | 7 - 8 | 931 | 80 | |
XM_011518340.2 | XP_011516642.1 | 8 - 9 | 1146 | 80 | |
XM_011518341.2 | XP_011516643.1 | 7 - 8 | 931 | 80 | |
XM_011518342.2 | XP_011516644.1 | 4 - 5 | 634 | 80 | |
XM_011518344.2 | XP_011516646.1 | 7 - 8 | 931 | 80 | |
XM_017014378.1 | XP_016869867.1 | 7 - 8 | 1129 | 80 | |
XM_017014379.1 | XP_016869868.1 | 7 - 8 | 1537 | 80 | |
XM_017014380.1 | XP_016869869.1 | 7 - 8 | 863 | 80 | |
XM_017014381.1 | XP_016869870.1 | 7 - 8 | 929 | 80 | |
XM_017014382.1 | XP_016869871.1 | 5 - 6 | 1394 | 80 | |
GenBank mRNA | AB055982.1 | - | 5 - 6 | 543 | 80 |
AB445477.1 | - | 5 - 6 | 543 | 80 | |
AF165281.1 | - | 4 - 5 | 483 | 80 | |
AF285167.1 | - | 6 - 7 | 833 | 80 | |
AJ012376.1 | - | 4 - 5 | 483 | 80 | |
AK024328.1 | - | 6 - 7 | 856 | 80 | |
BC146856.1 | - | 6 - 7 | 757 | 80 |