Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171968.1 | NP_001165439.1 | 8 - 9 | 1044 | 67 |
NM_021924.4 | NP_068743.2 | 8 - 9 | 1044 | 67 | |
NM_031264.3 | NP_112554.2 | 8 - 9 | 1044 | 67 | |
XM_006718253.3 | XP_006718316.1 | 8 - 9 | 944 | 67 | |
XM_011520188.2 | XP_011518490.1 | 8 - 9 | 944 | 67 | |
XM_011520189.2 | XP_011518491.1 | 8 - 9 | 944 | 67 | |
XM_011520190.2 | XP_011518492.1 | 8 - 9 | 944 | 67 | |
XM_011520191.2 | XP_011518493.1 | 8 - 9 | 944 | 67 | |
GenBank mRNA | AF258675.1 | - | 9 - 10 | 1200 | 67 |
AF258676.1 | - | 9 - 10 | 1200 | 67 | |
AF276242.1 | - | 8 - 9 | 879 | 67 | |
AF301909.1 | - | 8 - 9 | 879 | 67 | |
AK000226.1 | - | 8 - 9 | 1062 | 67 | |
AK000384.1 | - | 8 - 9 | 949 | 67 | |
AK300991.1 | - | 9 - 10 | 896 | 67 | |
AK308040.1 | - | 8 - 9 | 968 | 67 | |
AK309157.1 | - | 7 - 8 | 839 | 67 | |
AY358368.1 | - | 8 - 9 | 901 | 67 | |
BC038405.1 | - | 8 - 9 | 901 | 67 | |
BC107063.2 | - | 3 - 4 | 67 | ||
DQ491193.1 | - | 8 - 9 | 951 | 67 |