Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031264.3 | NP_112554.2 | 12 - 13 | 1545 | 87 |
GenBank mRNA | AF276242.1 | - | 12 - 13 | 1380 | 87 |
AK000384.1 | - | 12 - 13 | 1450 | 87 | |
AK300991.1 | - | 13 - 14 | 1397 | 87 | |
BC038405.1 | - | 12 - 13 | 1402 | 87 | |
DQ491193.1 | - | 12 - 13 | 1452 | 87 |