Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287223.1 | NP_001274152.1 | 8 - 9 | 991 | 120 |
NM_014139.2 | NP_054858.2 | 7 - 8 | 1160 | 120 | |
XM_011533321.2 | XP_011531623.1 | 4 - 5 | 385 | 120 | |
XM_017005647.1 | XP_016861136.1 | 12 - 13 | 1612 | 120 | |
XM_017005648.1 | XP_016861137.1 | 10 - 11 | 1291 | 120 | |
XM_017005649.1 | XP_016861138.1 | 11 - 12 | 1490 | 120 | |
XM_017005650.1 | XP_016861139.1 | 8 - 9 | 1291 | 120 | |
XM_017005651.1 | XP_016861140.1 | 8 - 9 | 817 | 120 | |
XM_017005652.1 | XP_016861141.1 | 10 - 11 | 1291 | 120 | |
GenBank mRNA | AF109737.1 | - | 7 - 8 | 1160 | 120 |
AF150882.1 | - | 7 - 8 | 1160 | 120 | |
AF188679.1 | - | 8 - 9 | 991 | 120 | |
AJ417790.1 | - | 7 - 8 | 961 | 120 |