Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017934.5 | NP_060404.3 | 24 - 25 | 3109 | 70 |
XM_005248729.4 | XP_005248786.1 | 24 - 25 | 3106 | 70 | |
XM_011535918.2 | XP_011534220.1 | 21 - 22 | 2765 | 70 | |
XM_011535919.1 | XP_011534221.1 | 24 - 25 | 3109 | 70 | |
XM_017010989.1 | XP_016866478.1 | 26 - 27 | 3251 | 70 | |
XM_017010990.1 | XP_016866479.1 | 23 - 24 | 2849 | 70 | |
GenBank mRNA | AF310250.1 | - | 1 - 2 | 116 | 70 |
AJ303102.1 | - | 24 - 25 | 2896 | 70 | |
AL161957.1 | - | 2 - 3 | 172 | 70 | |
BC137488.1 | - | 24 - 25 | 2989 | 70 | |
DQ924532.1 | - | 24 - 25 | 2883 | 70 |