Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182628.2 | NP_872434.2 | 12 - 13 | 1238 | 120 |
XM_006713623.2 | XP_006713686.1 | 13 - 14 | 1389 | 120 | |
XM_017006322.1 | XP_016861811.1 | 13 - 14 | 1381 | 120 | |
XM_017006323.1 | XP_016861812.1 | 13 - 14 | 1381 | 120 | |
XM_017006324.1 | XP_016861813.1 | 13 - 14 | 1381 | 120 | |
GenBank mRNA | AK097402.1 | - | 12 - 13 | 1238 | 120 |
BC101366.1 | - | 11 - 12 | 1171 | 120 | |
BC101367.2 | - | 11 - 12 | 1168 | 120 | |
BC101368.1 | - | 11 - 12 | 1168 | 120 | |
BC101369.1 | - | 11 - 12 | 1168 | 120 |