Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270.2 | NP_001261.2 | 10 - 11 | 1642 | 107 |
XM_005271866.4 | XP_005271923.1 | 11 - 12 | 2118 | 107 | |
XM_005271867.4 | XP_005271924.1 | 11 - 12 | 2118 | 107 | |
XM_011543112.2 | XP_011541414.1 | 11 - 12 | 2118 | 107 | |
GenBank mRNA | AF006513.1 | - | 10 - 11 | 1657 | 107 |
BC117134.1 | - | 10 - 11 | 1632 | 107 |