Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270.2 | NP_001261.2 | 34 - 35 | 4929 | 128 |
XM_005271866.4 | XP_005271923.1 | 36 - 37 | 5669 | 128 | |
XM_005271867.4 | XP_005271924.1 | 35 - 36 | 5405 | 128 | |
XM_017008991.1 | XP_016864480.1 | 20 - 21 | 2729 | 128 | |
XM_017008992.1 | XP_016864481.1 | 20 - 21 | 2574 | 128 | |
GenBank mRNA | AF006513.1 | - | 34 - 35 | 4944 | 128 |
AK094871.1 | - | 3 - 4 | 969 | 128 | |
BC117134.1 | - | 34 - 35 | 4919 | 128 | |
BQ436172.1 | - | 6 - 7 | 693 | 128 | |
BX648767.1 | - | 9 - 10 | 1447 | 128 |