Assay Design



Gene Transcripts

Gene Symbol
SENP1
Entrez Gene ID
29843
Gene Name
SUMO1/sentrin specific peptidase 1
Gene Aliases
SuPr-2
Chromosome Location
Chr.12: 48042893 - 48106308 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267594.1 NP_001254523.1 3 - 4 457 59
NM_001267595.1 NP_001254524.1 3 - 4 712 59
NR_051991.1 3 - 4 767 59
NR_051992.1 3 - 4 780 59
XM_006719361.3 XP_006719424.1 3 - 4 592 59
XM_006719362.2 XP_006719425.1 3 - 4 233 59
XM_011538244.2 XP_011536546.1 3 - 4 632 59
XM_011538245.2 XP_011536547.1 2 - 3 544 59
XM_017019229.1 XP_016874718.1 4 - 5 730 59
XM_017019230.1 XP_016874719.1 4 - 5 685 59
XM_017019231.1 XP_016874720.1 4 - 5 630 59
XM_017019232.1 XP_016874721.1 4 - 5 357 59
XM_017019233.1 XP_016874722.1 3 - 4 640 59
XM_017019234.1 XP_016874723.1 3 - 4 550 59
XM_017019235.1 XP_016874724.1 3 - 4 283 59
XM_017019236.1 XP_016874725.1 3 - 4 557 59
XM_017019237.1 XP_016874726.1 3 - 4 457 59
XM_017019238.1 XP_016874727.1 3 - 4 537 59
XM_017019239.1 XP_016874728.1 2 - 3 577 59
XM_017019240.1 XP_016874729.1 3 - 4 537 59
GenBank mRNA AF149770.1 - 1 - 2 132 59
AK292060.1 - 3 - 4 228 59
AK312874.1 - 3 - 4 457 59
AL582855.3 - 3 - 4 579 59
BC045639.1 - 3 - 4 528 59
BX537920.1 - 3 - 4 780 59
BX640784.1 - 3 - 4 611 59

View Full Product Details