Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267594.1 | NP_001254523.1 | 11 - 12 | 1446 | 117 |
NM_001267595.1 | NP_001254524.1 | 11 - 12 | 1701 | 117 | |
NR_051991.1 | 12 - 13 | 1887 | 117 | ||
NR_051992.1 | 12 - 13 | 1900 | 117 | ||
XM_006719361.3 | XP_006719424.1 | 11 - 12 | 1581 | 117 | |
XM_006719362.2 | XP_006719425.1 | 12 - 13 | 1353 | 117 | |
XM_011538244.2 | XP_011536546.1 | 10 - 11 | 1582 | 117 | |
XM_011538245.2 | XP_011536547.1 | 10 - 11 | 1533 | 117 | |
XM_017019229.1 | XP_016874718.1 | 12 - 13 | 1719 | 117 | |
XM_017019230.1 | XP_016874719.1 | 11 - 12 | 1635 | 117 | |
XM_017019231.1 | XP_016874720.1 | 12 - 13 | 1619 | 117 | |
XM_017019232.1 | XP_016874721.1 | 12 - 13 | 1346 | 117 | |
XM_017019233.1 | XP_016874722.1 | 11 - 12 | 1629 | 117 | |
XM_017019234.1 | XP_016874723.1 | 11 - 12 | 1539 | 117 | |
XM_017019235.1 | XP_016874724.1 | 11 - 12 | 1272 | 117 | |
XM_017019236.1 | XP_016874725.1 | 10 - 11 | 1507 | 117 | |
XM_017019237.1 | XP_016874726.1 | 10 - 11 | 1407 | 117 | |
XM_017019238.1 | XP_016874727.1 | 12 - 13 | 1657 | 117 | |
XM_017019239.1 | XP_016874728.1 | 11 - 12 | 1697 | 117 | |
XM_017019240.1 | XP_016874729.1 | 11 - 12 | 1618 | 117 | |
GenBank mRNA | AF149770.1 | - | 9 - 10 | 1121 | 117 |
AK292060.1 | - | 11 - 12 | 1217 | 117 | |
AK312874.1 | - | 11 - 12 | 1446 | 117 | |
BC045639.1 | - | 11 - 12 | 1517 | 117 | |
BX537920.1 | - | 12 - 13 | 1900 | 117 | |
BX640784.1 | - | 12 - 13 | 1731 | 117 |