Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004170.5 | NP_004161.4 | 4 - 5 | 696 | 81 |
XM_011518007.1 | XP_011516309.1 | 5 - 6 | 716 | 81 | |
XM_011518008.2 | XP_011516310.1 | 5 - 6 | 7961 | 81 | |
XM_011518009.2 | XP_011516311.1 | 4 - 5 | 7891 | 81 | |
XM_011518010.1 | XP_011516312.1 | 3 - 4 | 506 | 81 | |
XM_017015042.1 | XP_016870531.1 | 5 - 6 | 716 | 81 | |
XM_017015043.1 | XP_016870532.1 | 4 - 5 | 657 | 81 | |
GenBank mRNA | AB008536.1 | - | 4 - 5 | 634 | 81 |
AK313013.1 | - | 4 - 5 | 597 | 81 | |
BC033040.1 | - | 4 - 5 | 682 | 81 | |
U03506.1 | - | 4 - 5 | 461 | 81 | |
U06469.1 | - | 4 - 5 | 482 | 81 | |
U08989.1 | - | 4 - 5 | 608 | 81 |