Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286779.1 | NP_001273708.1 | 15 - 16 | 1513 | 110 |
NM_032293.4 | NP_115669.3 | 14 - 15 | 1650 | 110 | |
NR_104591.1 | 14 - 15 | 1650 | 110 | ||
XM_005252267.3 | XP_005252324.1 | 16 - 17 | 1505 | 110 | |
XM_005252268.2 | XP_005252325.1 | 16 - 17 | 1567 | 110 | |
XM_011519086.2 | XP_011517388.1 | 17 - 18 | 1577 | 110 | |
XM_011519087.2 | XP_011517389.1 | 17 - 18 | 1560 | 110 | |
XM_011519089.2 | XP_011517391.1 | 16 - 17 | 1463 | 110 | |
XM_011519090.2 | XP_011517392.1 | 15 - 16 | 1346 | 110 | |
XM_017015202.1 | XP_016870691.1 | 15 - 16 | 1543 | 110 | |
GenBank mRNA | AK095679.1 | - | 12 - 13 | 1422 | 110 |
AK293732.1 | - | 14 - 15 | 1548 | 110 | |
AK294970.1 | - | 14 - 15 | 1152 | 110 | |
AK296246.1 | - | 15 - 16 | 1513 | 110 | |
AL136573.1 | - | 14 - 15 | 1204 | 110 | |
BC034983.2 | - | 14 - 15 | 1260 | 110 |