Assay Design



Gene Transcripts

Gene Symbol
GARNL3
Entrez Gene ID
84253
Gene Name
GTPase activating Rap/RanGAP domain like 3
Gene Aliases
bA356B19.1
Chromosome Location
Chr.9: 127224471 - 127393655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286779.1 NP_001273708.1 28 - 29 3132 90
NM_032293.4 NP_115669.3 27 - 28 3269 90
NR_104591.1 26 - 27 3130 90
XM_005252267.3 XP_005252324.1 29 - 30 3124 90
XM_005252268.2 XP_005252325.1 29 - 30 3186 90
XM_011519086.2 XP_011517388.1 30 - 31 3196 90
XM_011519087.2 XP_011517389.1 30 - 31 3179 90
XM_011519089.2 XP_011517391.1 29 - 30 3082 90
XM_011519090.2 XP_011517392.1 28 - 29 2965 90
XM_017015202.1 XP_016870691.1 28 - 29 3162 90
GenBank mRNA AK126704.1 - 6 - 7 4357 90
AK293732.1 - 27 - 28 3167 90
AK294970.1 - 27 - 28 2771 90
AK296246.1 - 28 - 29 3132 90
AL136573.1 - 26 - 27 2684 90
AL834149.1 - 11 - 12 1401 90
BC034983.2 - 27 - 28 2879 90

View Full Product Details