Assay Design
Gene Transcripts
- Gene Symbol
- STRADA
- Entrez Gene ID
-
92335
- Gene Name
- STE20-related kinase adaptor alpha
- Gene Aliases
-
LYK5, NY-BR-96, PMSE, STRAD, Stlk
- Chromosome Location
-
Chr.17: 63702832 - 63741970 on Build GRCh38
View Full Product Details