Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324281.1 | NP_001311210.1 | 19 - 20 | 2974 | 85 |
NM_032415.5 | NP_115791.3 | 18 - 19 | 2913 | 85 | |
XM_011515586.2 | XP_011513888.1 | 19 - 20 | 2942 | 85 | |
XM_011515587.2 | XP_011513889.1 | 18 - 19 | 2905 | 85 | |
GenBank mRNA | AF322641.1 | - | 18 - 19 | 2815 | 85 |
AF352576.1 | - | 16 - 17 | 2488 | 85 | |
AK074049.1 | - | 18 - 19 | 2833 | 85 | |
BC111719.1 | - | 17 - 18 | 2599 | 85 |