Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024831.7 | NP_079107.6 | 1 - 2 | 484 | 99 |
XM_006716485.2 | XP_006716548.1 | 1 - 2 | 270 | 99 | |
XM_017014004.1 | XP_016869493.1 | 1 - 2 | 270 | 99 | |
GenBank mRNA | AF286340.1 | - | 1 - 2 | 574 | 99 |
AF432215.1 | - | 1 - 2 | 232 | 99 | |
AK314693.1 | - | 1 - 2 | 258 | 99 | |
AY028423.1 | - | 1 - 2 | 268 | 99 | |
BM853546.1 | - | 1 - 2 | 484 | 99 | |
DB082992.1 | - | 1 - 2 | 272 | 99 |