Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317902.1 | NP_001304831.1 | 5 - 6 | 1689 | 73 |
NM_024831.7 | NP_079107.6 | 6 - 7 | 1754 | 73 | |
XM_005251328.2 | XP_005251385.1 | 5 - 6 | 1469 | 73 | |
XM_006716485.2 | XP_006716548.1 | 6 - 7 | 1540 | 73 | |
XM_017014004.1 | XP_016869493.1 | 6 - 7 | 1540 | 73 | |
GenBank mRNA | AF286340.1 | - | 6 - 7 | 1844 | 73 |
AF432215.1 | - | 6 - 7 | 1502 | 73 | |
AK055178.1 | - | 3 - 4 | 628 | 73 | |
AK314693.1 | - | 6 - 7 | 1528 | 73 | |
AY028423.1 | - | 6 - 7 | 1538 | 73 | |
AY534911.1 | - | 3 - 4 | 544 | 73 | |
BX648548.1 | - | 5 - 6 | 1452 | 73 |