Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317902.1 | NP_001304831.1 | 6 - 7 | 1864 | 148 |
NM_024831.7 | NP_079107.6 | 7 - 8 | 1929 | 148 | |
XM_005251328.2 | XP_005251385.1 | 6 - 7 | 1644 | 148 | |
XM_006716485.2 | XP_006716548.1 | 7 - 8 | 1715 | 148 | |
XM_017014004.1 | XP_016869493.1 | 7 - 8 | 1715 | 148 | |
GenBank mRNA | AF286340.1 | - | 7 - 8 | 2019 | 148 |
AF432215.1 | - | 7 - 8 | 1677 | 148 | |
AK055178.1 | - | 4 - 5 | 803 | 148 | |
AK314693.1 | - | 7 - 8 | 1703 | 148 | |
AL832426.1 | - | 1 - 2 | 185 | 148 | |
AY028423.1 | - | 7 - 8 | 1713 | 148 | |
AY534911.1 | - | 4 - 5 | 719 | 148 | |
BX648548.1 | - | 6 - 7 | 1627 | 148 |