Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000721.3 | NP_000712.2 | 5 - 6 | 965 | 94 |
NM_001205293.1 | NP_001192222.1 | 5 - 6 | 965 | 94 | |
NM_001205294.1 | NP_001192223.1 | 5 - 6 | 965 | 94 | |
XM_017002243.1 | XP_016857732.1 | 7 - 8 | 1630 | 94 | |
XM_017002244.1 | XP_016857733.1 | 7 - 8 | 1631 | 94 | |
XM_017002245.1 | XP_016857734.1 | 7 - 8 | 1631 | 94 | |
XM_017002246.1 | XP_016857735.1 | 7 - 8 | 1631 | 94 | |
XM_017002247.1 | XP_016857736.1 | 7 - 8 | 1630 | 94 | |
XM_017002248.1 | XP_016857737.1 | 7 - 8 | 1631 | 94 | |
XM_017002249.1 | XP_016857738.1 | 7 - 8 | 1630 | 94 | |
XM_017002250.1 | XP_016857739.1 | 7 - 8 | 1630 | 94 | |
XM_017002251.1 | XP_016857740.1 | 7 - 8 | 1288 | 94 | |
GenBank mRNA | AK309070.1 | - | 5 - 6 | 1202 | 94 |
L27745.1 | - | 5 - 6 | 770 | 94 | |
L29384.1 | - | 5 - 6 | 935 | 94 | |
L29385.1 | - | 5 - 6 | 935 | 94 |