Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020318.2 | NP_064714.2 | 5 - 6 | 3596 | 67 |
XM_005245422.3 | XP_005245479.1 | 5 - 6 | 3643 | 67 | |
XM_011509857.1 | XP_011508159.1 | 6 - 7 | 3513 | 67 | |
XM_011509858.2 | XP_011508160.1 | 5 - 6 | 3856 | 67 | |
XM_017002023.1 | XP_016857512.1 | 6 - 7 | 3694 | 67 | |
XM_017002024.1 | XP_016857513.1 | 6 - 7 | 3741 | 67 | |
XM_017002025.1 | XP_016857514.1 | 5 - 6 | 3847 | 67 | |
GenBank mRNA | AF311940.1 | - | 4 - 5 | 2432 | 67 |
AF342989.1 | - | 5 - 6 | 3596 | 67 | |
AJ278348.1 | - | 4 - 5 | 2395 | 67 | |
BC152552.1 | - | 5 - 6 | 3584 | 67 |