Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142674.1 | NP_001136146.1 | 1 - 2 | 97 | 72 |
XM_005253073.4 | XP_005253130.1 | 1 - 2 | 81 | 72 | |
XM_017018168.1 | XP_016873657.1 | 1 - 2 | 80 | 72 | |
GenBank mRNA | BI818660.1 | - | 1 - 2 | 97 | 72 |