Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142674.1 | NP_001136146.1 | 8 - 9 | 857 | 59 |
NM_001142675.1 | NP_001136147.1 | 7 - 8 | 967 | 59 | |
NM_001142676.1 | NP_001136148.1 | 8 - 9 | 828 | 59 | |
NM_023947.3 | NP_076436.3 | 7 - 8 | 753 | 59 | |
XM_005253072.2 | XP_005253129.1 | 7 - 8 | 840 | 59 | |
XM_006718282.2 | XP_006718345.1 | 8 - 9 | 828 | 59 | |
XM_011520299.1 | XP_011518601.1 | 7 - 8 | 1064 | 59 | |
XM_011520301.2 | XP_011518603.1 | 7 - 8 | 1064 | 59 | |
XM_011520302.2 | XP_011518604.1 | 7 - 8 | 1064 | 59 | |
XM_017018166.1 | XP_016873655.1 | 8 - 9 | 914 | 59 | |
XM_017018167.1 | XP_016873656.1 | 7 - 8 | 719 | 59 | |
XM_017018168.1 | XP_016873657.1 | 8 - 9 | 840 | 59 | |
GenBank mRNA | AF212229.1 | - | 8 - 9 | 791 | 59 |
AK027711.1 | - | 7 - 8 | 702 | 59 | |
AK075413.1 | - | 8 - 9 | 764 | 59 | |
AK124697.1 | - | 7 - 8 | 1329 | 59 | |
AK225025.1 | - | 8 - 9 | 793 | 59 | |
AK225398.1 | - | 8 - 9 | 776 | 59 | |
AK301245.1 | - | 7 - 8 | 813 | 59 | |
BC000001.2 | - | 8 - 9 | 777 | 59 | |
BC013642.1 | - | 5 - 6 | 389 | 59 | |
BC095409.1 | - | 7 - 8 | 689 | 59 | |
BI821281.1 | - | 7 - 8 | 719 | 59 |