Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011545276.2 | XP_011543578.1 | 18 - 19 | 2549 | 63 |
XM_011545277.2 | XP_011543579.1 | 18 - 19 | 2937 | 63 | |
XM_011545278.2 | XP_011543580.1 | 18 - 19 | 3000 | 63 | |
XM_011545279.2 | XP_011543581.1 | 16 - 17 | 2519 | 63 | |
XM_011545280.1 | XP_011543582.1 | 15 - 16 | 1852 | 63 | |
XM_011545281.2 | XP_011543583.1 | 11 - 12 | 1672 | 63 | |
XM_011545286.1 | XP_011543588.1 | 11 - 12 | 1408 | 63 | |
GenBank mRNA | AK091658.1 | - | 1 - 2 | 1911 | 63 |