Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317895.1 | NP_001304824.1 | 3 - 4 | 914 | 80 |
NM_032789.4 | NP_116178.2 | 4 - 5 | 790 | 80 | |
NR_134234.1 | 4 - 5 | 790 | 80 | ||
XM_011517336.2 | XP_011515638.1 | 4 - 5 | 757 | 80 | |
GenBank mRNA | AK027370.1 | - | 4 - 5 | 747 | 80 |
AK128537.1 | - | 5 - 6 | 605 | 80 | |
AK222914.1 | - | 4 - 5 | 773 | 80 | |
AK303317.1 | - | 3 - 4 | 839 | 80 | |
AK308657.1 | - | 3 - 4 | 594 | 80 | |
AK309251.1 | - | 3 - 4 | 494 | 80 | |
BC020378.1 | - | 4 - 5 | 765 | 80 | |
BC030549.1 | - | 4 - 5 | 764 | 80 | |
BC132961.1 | - | 4 - 5 | 741 | 80 | |
BC136542.1 | - | 4 - 5 | 741 | 80 | |
BC144235.1 | - | 4 - 5 | 741 | 80 |