Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160372.3 | NP_001153844.1 | 12 - 13 | 1991 | 74 |
NM_001321646.1 | NP_001308575.1 | 11 - 12 | 1964 | 74 | |
NM_031466.7 | NP_113654.4 | 12 - 13 | 2166 | 74 | |
XM_011517326.2 | XP_011515628.1 | 11 - 12 | 2139 | 74 | |
XM_011517328.2 | XP_011515630.1 | 12 - 13 | 2166 | 74 | |
XM_011517330.2 | XP_011515632.1 | 4 - 5 | 531 | 74 | |
XM_017013893.1 | XP_016869382.1 | 12 - 13 | 2166 | 74 | |
XM_017013894.1 | XP_016869383.1 | 5 - 6 | 867 | 74 | |
GenBank mRNA | AB067469.1 | - | 12 - 13 | 1941 | 74 |
AK026394.1 | - | 12 - 13 | 1983 | 74 | |
AK027689.1 | - | 1 - 2 | 91 | 74 | |
AK128755.1 | - | 12 - 13 | 2167 | 74 | |
AK308815.1 | - | 1 - 2 | 262 | 74 | |
AL833973.1 | - | 10 - 11 | 1266 | 74 | |
AY190606.1 | - | 10 - 11 | 1245 | 74 | |
BC006206.2 | - | 10 - 11 | 1387 | 74 |