Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160372.3 | NP_001153844.1 | 21 - 22 | 3192 | 100 |
NM_001321646.1 | NP_001308575.1 | 20 - 21 | 3165 | 100 | |
NM_031466.7 | NP_113654.4 | 21 - 22 | 3367 | 100 | |
XM_011517326.2 | XP_011515628.1 | 20 - 21 | 3340 | 100 | |
XM_011517330.2 | XP_011515632.1 | 13 - 14 | 1732 | 100 | |
XM_017013894.1 | XP_016869383.1 | 14 - 15 | 2068 | 100 | |
GenBank mRNA | AB067469.1 | - | 21 - 22 | 3142 | 100 |
AK026394.1 | - | 21 - 22 | 3184 | 100 | |
AK027689.1 | - | 10 - 11 | 1292 | 100 | |
AK128755.1 | - | 21 - 22 | 3368 | 100 | |
AK308815.1 | - | 10 - 11 | 1463 | 100 | |
AL833973.1 | - | 19 - 20 | 2467 | 100 | |
AY190606.1 | - | 19 - 20 | 2446 | 100 | |
AY630619.1 | - | 2 - 3 | 247 | 100 | |
BC003650.2 | - | 7 - 8 | 911 | 100 | |
BC006206.2 | - | 19 - 20 | 2588 | 100 | |
BC065288.1 | - | 4 - 5 | 452 | 100 |