Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160372.3 | NP_001153844.1 | 22 - 23 | 3419 | 89 |
NM_001321646.1 | NP_001308575.1 | 21 - 22 | 3392 | 89 | |
NM_031466.7 | NP_113654.4 | 22 - 23 | 3594 | 89 | |
XM_011517326.2 | XP_011515628.1 | 21 - 22 | 3567 | 89 | |
XM_011517330.2 | XP_011515632.1 | 14 - 15 | 1959 | 89 | |
XM_017013894.1 | XP_016869383.1 | 15 - 16 | 2295 | 89 | |
GenBank mRNA | AB067469.1 | - | 22 - 23 | 3369 | 89 |
AK026394.1 | - | 22 - 23 | 3411 | 89 | |
AK027689.1 | - | 11 - 12 | 1519 | 89 | |
AK128755.1 | - | 22 - 23 | 3595 | 89 | |
AK308815.1 | - | 11 - 12 | 1690 | 89 | |
AL833973.1 | - | 20 - 21 | 2694 | 89 | |
AY190606.1 | - | 20 - 21 | 2673 | 89 | |
AY630619.1 | - | 3 - 4 | 474 | 89 | |
BC003650.2 | - | 8 - 9 | 1138 | 89 | |
BC006206.2 | - | 20 - 21 | 2815 | 89 | |
BC065288.1 | - | 5 - 6 | 679 | 89 |