Assay Design



Gene Transcripts

Gene Symbol
TRAPPC9
Entrez Gene ID
83696
Gene Name
trafficking protein particle complex 9
Gene Aliases
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120
Chromosome Location
Chr.8: 139727725 - 140458579 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160372.3 NP_001153844.1 8 - 9 1491 77
NM_001321646.1 NP_001308575.1 7 - 8 1464 77
NM_031466.7 NP_113654.4 8 - 9 1666 77
XM_011517326.2 XP_011515628.1 7 - 8 1639 77
XM_011517328.2 XP_011515630.1 8 - 9 1666 77
XM_017013893.1 XP_016869382.1 8 - 9 1666 77
GenBank mRNA AB067469.1 - 8 - 9 1441 77
AK026394.1 - 8 - 9 1483 77
AK128755.1 - 8 - 9 1667 77
AL833973.1 - 6 - 7 767 77
AY190606.1 - 6 - 7 745 77
BC006206.2 - 6 - 7 887 77

View Full Product Details