Assay Design



Gene Transcripts

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
Chromosome Location
Chr.1: 28887091 - 29120046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166005.1 NP_001159477.1 17 - 18 2440 65
NM_001166007.1 NP_001159479.1 14 - 15 2261 65
NM_203342.2 NP_976217.1 17 - 18 2478 65
NM_203343.2 NP_976218.1 13 - 14 2173 65
XM_005245753.1 XP_005245810.1 18 - 19 2965 65
XM_005245757.1 XP_005245814.1 16 - 17 2860 65
XM_005245760.1 XP_005245817.1 18 - 19 2566 65
XM_005245761.1 XP_005245818.1 17 - 18 2515 65
XM_005245763.1 XP_005245820.1 16 - 17 2473 65
XM_005245764.1 XP_005245821.1 15 - 16 2416 65
XM_005245765.1 XP_005245822.1 15 - 16 2410 65
XM_005245769.1 XP_005245826.1 14 - 15 2353 65
XM_005245770.1 XP_005245827.1 14 - 15 2350 65
XM_006710434.1 XP_006710497.1 19 - 20 3001 65
XM_006710439.1 XP_006710502.1 18 - 19 2551 65
XM_011540956.1 XP_011539258.1 19 - 20 3016 65
XM_011540957.1 XP_011539259.1 19 - 20 3013 65
XM_011540958.1 XP_011539260.1 19 - 20 2977 65
XM_011540959.1 XP_011539261.1 18 - 19 2974 65
XM_011540960.1 XP_011539262.1 17 - 18 2917 65
XM_011540961.1 XP_011539263.1 17 - 18 2911 65
XM_011540962.1 XP_011539264.1 16 - 17 2854 65
XM_011540964.1 XP_011539266.1 19 - 20 2912 65
XM_017000581.1 XP_016856070.1 18 - 19 2911 65
XM_017000582.1 XP_016856071.1 18 - 19 2908 65
XM_017000583.1 XP_016856072.1 16 - 17 2851 65
XM_017000584.1 XP_016856073.1 19 - 20 3307 65
XM_017000585.1 XP_016856074.1 16 - 17 3142 65
XM_017000586.1 XP_016856075.1 16 - 17 2446 65
XM_017000589.1 XP_016856078.1 13 - 14 2248 65
XM_017000590.1 XP_016856079.1 13 - 14 2245 65
XM_017000593.1 XP_016856082.1 19 - 20 2909 65
XM_017000594.1 XP_016856083.1 17 - 18 2813 65
XM_017000595.1 XP_016856084.1 17 - 18 2807 65
XM_017000596.1 XP_016856085.1 16 - 17 2750 65
XM_017000597.1 XP_016856086.1 16 - 17 2747 65
GenBank mRNA AB209649.1 - 14 - 15 2261 65
AF156225.1 - 16 - 17 2313 65
AK299093.1 - 2 - 3 320 65
AK307816.1 - 2 - 3 356 65
AL833483.1 - 1 - 2 1708 65
BC096104.1 - 11 - 12 1567 65
BC096105.1 - 14 - 15 1948 65
BC114450.1 - 14 - 15 1948 65
J03796.1 - 13 - 14 2092 65
M61733.1 - 17 - 18 2459 65

View Full Product Details