Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012392.3 | NP_036524.1 | 3 - 4 | 850 | 80 |
NR_033686.1 | 2 - 3 | 549 | 80 | ||
NR_033688.1 | 3 - 4 | 691 | 80 | ||
XM_011541745.1 | XP_011540047.1 | 3 - 4 | 477 | 80 | |
XM_011541746.2 | XP_011540048.1 | 3 - 4 | 944 | 80 | |
XM_011541747.1 | XP_011540049.1 | 3 - 4 | 819 | 80 | |
XM_017001680.1 | XP_016857169.1 | 3 - 4 | 808 | 80 | |
XM_017001681.1 | XP_016857170.1 | 3 - 4 | 2115 | 80 | |
GenBank mRNA | AB018357.1 | - | 3 - 4 | 489 | 80 |
AB026628.1 | - | 3 - 4 | 489 | 80 | |
AK001420.1 | - | 3 - 4 | 502 | 80 | |
AY359011.1 | - | 3 - 4 | 493 | 80 | |
BC002773.2 | - | 3 - 4 | 479 | 80 | |
BC012561.1 | - | 3 - 4 | 495 | 80 | |
BG546473.1 | - | 2 - 3 | 209 | 80 | |
CR542139.1 | - | 3 - 4 | 477 | 80 |