Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289135.2 | NP_001276064.1 | 31 - 32 | 5459 | 66 |
NM_001289136.2 | NP_001276065.1 | 31 - 32 | 5447 | 66 | |
NM_004787.3 | NP_004778.1 | 32 - 33 | 5471 | 66 | |
XM_005248211.3 | XP_005248268.1 | 32 - 33 | 5483 | 66 | |
XM_006713986.3 | XP_006714049.1 | 31 - 32 | 5411 | 66 | |
XM_011513909.2 | XP_011512211.1 | 32 - 33 | 3511 | 66 | |
XM_011513910.2 | XP_011512212.2 | 20 - 21 | 4363 | 66 | |
XM_017008845.1 | XP_016864334.1 | 32 - 33 | 3498 | 66 | |
GenBank mRNA | AB017168.1 | - | 32 - 33 | 3764 | 66 |
AF055585.1 | - | 31 - 32 | 4092 | 66 | |
AF133270.1 | - | 31 - 32 | 3548 | 66 | |
AK027326.1 | - | 4 - 5 | 534 | 66 | |
BC117190.1 | - | 32 - 33 | 3639 | 66 | |
BC143978.1 | - | 31 - 32 | 3615 | 66 |