Assay Design



Gene Transcripts

Gene Symbol
SLIT2
Entrez Gene ID
9353
Gene Name
slit guidance ligand 2
Gene Aliases
SLIL3, Slit-2
Chromosome Location
Chr.4: 20251905 - 20620561 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289135.2 NP_001276064.1 6 - 7 2450 116
NM_001289136.2 NP_001276065.1 6 - 7 2450 116
NM_004787.3 NP_004778.1 6 - 7 2450 116
XM_005248211.3 XP_005248268.1 6 - 7 2450 116
XM_006713986.3 XP_006714049.1 6 - 7 2450 116
XM_011513909.2 XP_011512211.1 6 - 7 478 116
XM_017008845.1 XP_016864334.1 6 - 7 477 116
GenBank mRNA AB017168.1 - 6 - 7 743 116
AF055585.1 - 6 - 7 1095 116
AF133270.1 - 6 - 7 539 116
BC117190.1 - 6 - 7 618 116
BC143978.1 - 6 - 7 618 116

View Full Product Details