Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005349.3 | NP_005340.2 | 3 - 4 | 330 | 73 |
NM_015874.4 | NP_056958.3 | 2 - 3 | 277 | 73 | |
NM_203283.2 | NP_976028.1 | 2 - 3 | 214 | 73 | |
NM_203284.2 | NP_976029.1 | 3 - 4 | 460 | 73 | |
XM_005248161.3 | XP_005248218.1 | 3 - 4 | 344 | 73 | |
XM_011513840.2 | XP_011512142.1 | 3 - 4 | 736 | 73 | |
XM_017008170.1 | XP_016863659.1 | 2 - 3 | 133 | 73 | |
XM_017008171.1 | XP_016863660.1 | 2 - 3 | 112 | 73 | |
XM_017008172.1 | XP_016863661.1 | 3 - 4 | 275 | 73 | |
XM_017008173.1 | XP_016863662.1 | 3 - 4 | 553 | 73 | |
XM_017008174.1 | XP_016863663.1 | 3 - 4 | 369 | 73 | |
XM_017008175.1 | XP_016863664.1 | 3 - 4 | 285 | 73 | |
GenBank mRNA | AK302230.1 | - | 2 - 3 | 67 | 73 |
AK303244.1 | - | 2 - 3 | 126 | 73 | |
AK311419.1 | - | 2 - 3 | 113 | 73 | |
BC020780.1 | - | 2 - 3 | 214 | 73 | |
BC064976.1 | - | 3 - 4 | 235 | 73 | |
BE269143.1 | - | 3 - 4 | 240 | 73 | |
BM472788.1 | - | - | 284 | 73 | |
BM555733.1 | - | 3 - 4 | 438 | 73 | |
BQ216033.1 | - | 2 - 3 | 258 | 73 | |
DC361185.1 | - | 2 - 3 | 277 | 73 | |
DC389691.1 | - | 3 - 4 | 460 | 73 | |
L07872.1 | - | 2 - 3 | 172 | 73 | |
L07874.1 | - | 2 - 3 | 289 | 73 | |
L07875.1 | - | 3 - 4 | 352 | 73 | |
L07876.1 | - | 2 - 3 | 217 | 73 |